Orders LinkContactUsLinkComponent
Phase separating paper, grade 480
Phase separating paper, grade 480
   Sustainable Options Available
Supplier:   Sartorius
Phase separating paper, grade 480
Supplier:  Sartorius
   Sustainable Options Available
Certificates
Phase separating paper, grade 480
8 Options Available Below
Product Details & Documents
This phase separating paper is impregnated with stabilised silicon, thus rendering it hydrophobic and it retains water, but allows solvents to flow through. The flow stops automatically when the entire solvent has passed through, this eliminates the need to use separating funnels.
Used for filtration of organic solvents that have been contaminated with water to separate the aqueous from the organic phase, filtration of extracting solvents in clinical or medical labs and separation of emulsions that are formed during the extraction of aqueous plant or drug solutions.
Specifications for Product Family
Specifications
  • Catalog No:
  • SARTFT-3-602-090
  • SARTFT-3-602-110
  • SARTFT-3-602-125
  • SARTFT-3-602-150
  • SARTFT-3-602-185
  • SARTFT-4-602-125
  • SARTFT-4-602-150
  • SARTFT-4-602-185
  • Weight:
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • Thickness:
  • 0,19 mm
  • 0,19 mm
  • 0,19 mm
  • 0,19 mm
  • 0,19 mm
  • 0,19 mm
  • 0,19 mm
  • 0,19 mm
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
Specifications
Product Family Options
Filter discs
Product Information
  • Grade
    Size
    Availability
    Price
  • 480
    90 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 25,70 €
    25,70 €
    1 * 100 ST
       25,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-3-602-090
    Weight
    85 g/m²
    Thickness
    0,19 mm
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 480
    110 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 37,30 €
    37,30 €
    1 * 100 ST
       37,30 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-3-602-110
    Weight
    85 g/m²
    Thickness
    0,19 mm
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 480
    125 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 54,10 €
    54,10 €
    1 * 100 ST
       54,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-3-602-125
    Weight
    85 g/m²
    Thickness
    0,19 mm
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 480
    150 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 84,80 €
    84,80 €
    1 * 100 ST
       84,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-3-602-150
    Weight
    85 g/m²
    Thickness
    0,19 mm
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 480
    185 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 94,00 €
    94,00 €
    1 * 100 ST
       94,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-3-602-185
    Weight
    85 g/m²
    Thickness
    0,19 mm
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
Folded filters
Product Information
  • Grade
    Size
    Availability
    Price
  • 480
    125 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 113,00 €
    113,00 €
    1 * 100 ST
       113,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-4-602-125
    Weight
    85 g/m²
    Thickness
    0,19 mm
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 480
    150 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 113,00 €
    113,00 €
    1 * 100 ST
       113,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-4-602-150
    Weight
    85 g/m²
    Thickness
    0,19 mm
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 480
    185 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 146,00 €
    146,00 €
    1 * 100 ST
       146,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SARTFT-4-602-185
    Weight
    85 g/m²
    Thickness
    0,19 mm
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact