Zinc
Plastic bottle for solids, 250 g
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
- CAS Number:7440-66-6
- Formula:Zn
- MDL Number:MFCD00011291
- Molecular Weight:65.39 g/mol
- Merck Index:12,10255
- Boiling Pt:908 °C (1013 hPa)
- Melting Pt:420 °C
- Density:7.14 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-175-3
Documents
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



