Order Entry
Belgium
Orders LinkContactUsLinkComponent
Zinc

Zinc

Plastic bottle for solids, 500 g
Catalog # 29071.261
Supplier:   VWR Chemicals
Zinc
Catalog # 29071.261
Supplier:  VWR Chemicals
CAS Number:  7440-66-6
Zinc
Zinc

Plastic bottle for solids, 500 g

CAS Number:  7440-66-6
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:7440-66-6
  • Formula:Zn
  • MDL Number:MFCD00011291
  • Molecular Weight:65.39 g/mol
  • Merck Index:12,10255
  • Boiling Pt:908 °C (1013 hPa)
  • Melting Pt:420 °C
  • Density:7.14 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-175-3

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.