ContactUsLinkComponent
Whatman™ 903 Proteinsaver Sample Collection Card
Whatman™ 903 Proteinsaver Sample Collection Card
Catalog # WHAT10531018
undefined
Whatman™ 903 Proteinsaver Sample Collection Card
Catalog # WHAT10531018
Specifications
  • Description:
    Proteinsaver cards (EU)
  • Environmentally Preferable:
  • Pk:
    100
Specifications
About this item
Whatman 903 sample collection cards are designed for the collection of biological fluids.
  • Ready to use cards saving time during sample collection, transport, and archiving
  • Strict quality control and ISO 9001 manufacturing standards ensure high quality, reproducibility, and purity
  • Choice of five-spot Proteinsaver card or four-spot Proteinsaver snap-apart card
The sample collection area contains five circles, each circle holds 75 to 80 µl of sample. Wraparound cover has spaces for name and date of collection and is imprinted with the biohazard symbol. The Proteinsaver card fits into Whatman™ foil barrier resealable bags for storage (available separately).
The cards are ready to use and provide easy collection, identification, and transport of samples, without the need for costly freezers or dry ice delivery.
Certifications: Under current regulations, specimen collection cards, including neonatal screening cards, which are to be used for human diagnostic tests, are classified as "other" IVD devices under the European IVD Directive and require the CE mark if sold within the European Union. All 903 specimen collection devices manufactured and printed by Whatman for neonatal testing in the EU undergo post-printing quality control and carry the CE mark.
Caution: Protein saver cards are for research use only. Whatman will assume no responsibility for the quality or performance of 903 collection devices converted, printed, or packaged by third party suppliers.