Cleanroom boot covers, Kimtech™ A5
About this item
Sterile Kimtech™ A5 cleanroom boot covers are made of SMS (Spunbond Melt-blown Spunbond) material that gives them a cloth-like feeling. Available with standard vinyl sole or with wrap-around vinyl foot.
- Wrap-around vinyl foot is seamed above the floor line to reduce risk of liquid penetration and trip hazards
- Tunnelled elastic and bound seams
- Two extra-long ties
Suitable for use in Class 100, ISO 5, M3.5 environments.
*One size fits all
: PPE Cat I (EU) 2016/425
: Vacuum packed in pairs for added sterility assurance.
4 Options Available Below
Product Details & Documents
Sterile Kimtech™ A5 cleanroom boot covers are made of SMS (Spunbond Melt-blown Spunbond) material that gives them a cloth-like feeling. Available with standard vinyl sole or with wrap-around vinyl foot.
- Wrap-around vinyl foot is seamed above the floor line to reduce risk of liquid penetration and trip hazards
- Tunnelled elastic and bound seams
- Two extra-long ties
Suitable for use in Class 100, ISO 5, M3.5 environments.
*One size fits all
: PPE Cat I (EU) 2016/425
: Vacuum packed in pairs for added sterility assurance.
Specifications for Product Family
Specifications
- Catalog No:
- 113-7736
- 113-7735
- 113-1227
- 113-8197
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
Specifications
Product Family Options
Standard vinyl sole
Product Information
- ColourShoe sizeSterileAvailabilityPrice
- 113-7736Standard vinyl soleSpecifications:
Waste Reducing More
Specifications
Cat. No.113-7736Environmentally PreferableWaste ReducingCleanroom class100/ISO 5Garment applicationCleanroomWaste Reducing - 113-7735Standard vinyl soleSpecifications:
Waste Reducing More
Specifications
Cat. No.113-7735Environmentally PreferableWaste ReducingCleanroom class100/ISO 5Garment applicationCleanroomWaste Reducing
Wrap-around vinyl foot
Product Information
- ColourShoe sizeSterileAvailabilityPrice
- 113-1227Wrap-around vinyl footSpecifications:
Waste Reducing More
Specifications
Cat. No.113-1227Environmentally PreferableWaste ReducingCleanroom class100/ISO 5Garment applicationCleanroomWaste Reducing - 113-8197Wrap-around vinyl footSpecifications:
Waste Reducing More
Specifications
Cat. No.113-8197Environmentally PreferableWaste ReducingCleanroom class100/ISO 5Garment applicationCleanroomWaste Reducing
Complementary Products
Product Information
- Catalog #DescriptionSterileClothing SizeColourLengthAvailabilityPrice
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.