Vacuum filtering flasks, Büchner, with side arm socket
Supplier: Brand
Jump to:
About this item
Borosilicate glass 3.3.
- Erlenmeyer shape
- Ground socket to accommodate vacuum tubing of 15 to 18 mm external diameter (e.g. 6×5 or 8×5 mm)
- Fulfil the regulations of the 'Equipment and product safety regulations'
For vacuum use. Filtration flasks with side arm socket have not only made work in preparation and analytical laboratories easier and simpler, but have also reduced accident risk.
Certifications: DIN 12476 and ISO 6556
4 Options Available Below
Product Details & Documents
Borosilicate glass 3.3.
- Erlenmeyer shape
- Ground socket to accommodate vacuum tubing of 15 to 18 mm external diameter (e.g. 6×5 or 8×5 mm)
- Fulfil the regulations of the 'Equipment and product safety regulations'
For vacuum use. Filtration flasks with side arm socket have not only made work in preparation and analytical laboratories easier and simpler, but have also reduced accident risk.
Certifications: DIN 12476 and ISO 6556
Product Family Options
Product Information
- CapacityHeightExt.ØNeck I-ØAvailabilityPrice
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.