Mailing containers
Supplier: DAKLAPACK
Jump to:
About this item
Solid PP.
- Transport blisters are designed for sending medical and biological samples
- Can be sent as letter, saving significant postage costs
Used in combination with the liquid absorbent insert and SafetyBag, they meet the packaging requirements of most international postal services.
5 Options Available Below
Product Details & Documents
Solid PP.
- Transport blisters are designed for sending medical and biological samples
- Can be sent as letter, saving significant postage costs
Used in combination with the liquid absorbent insert and SafetyBag, they meet the packaging requirements of most international postal services.
Product Family Options
Product Information
- AvailabilityPrice
- 216-0802Stool sample transport blister, one position
- DAKL460168Blood transport blister, one position
- 216-0790Blood transport blister, two positions
- 216-0800Blood transport blister, three positions
- 216-0801Blood transport blister, six positions
Accessories
Product Information
- Catalog #AvailabilityPrice
- Liquidproof safety bag, large format, PE, 255×385 mmLiquidproof safety bag, large format, PE, 255×385 mmDAKL460905Log in for availability1 * 500 ST - 1 440,00 €
Product Information
- Catalog #AvailabilityPrice
- Liquidproof safety bag, for use with POLYMED, PE, international text, 165×170 mmLiquidproof safety bag, for use with POLYMED, PE, international text, 165×170 mmDAKL460909Log in for availability1 * 100 ST - 99,50 €
Product Information
- Catalog #AvailabilityPrice
- Liquidproof safety bag, large format, PE, 385×580 mmLiquidproof safety bag, large format, PE, 385×580 mmDAKL460910Log in for availability1 * 100 ST - 377,00 €
Product Information
- Catalog #AvailabilityPrice
- Liquidproof safety bag, large format, PE, 590×620 mmLiquidproof safety bag, large format, PE, 590×620 mmDAKL460911Log in for availability1 * 100 ST - 526,00 €
Product Information
- Catalog #AvailabilityPrice
- Liquidproof safety bag, PE, for use with POLYMED, international text, 165×265 mmLiquidproof safety bag, PE, for use with POLYMED, international text, 165×265 mmDAKL460915Log in for availability1 * 500 ST - 588,00 €
Product Information
- Catalog #AvailabilityPrice
- Liquidproof safety bag for use with POLYMED, PE, dutch text, 165×275 mmLiquidproof safety bag for use with POLYMED, PE, dutch text, 165×275 mm216-0789Log in for availability1 * 500 ST - 560,00 €
Product Information
- Catalog #AvailabilityPrice
- Liquidproof safety bag with extra pocket, PE
Product Information
- Catalog #AvailabilityPrice
- Absorbing insert sheet, 90×127 mm
Product Information
- Catalog #AvailabilityPrice
- Multi-swab blister, six positions