Orders LinkContactUsLinkComponent
Dropping bottles, PTFE
Dropping bottles, PTFE
Supplier:   Brand
Dropping bottles, PTFE
Supplier:  Brand
Certificates
Dropping bottles, PTFE
Dropping bottles, PTFE
2 Options Available Below
Product Details & Documents
PTFE, white.
  • Flexible wall for easy use
  • Screw fit cap
  • Guaranteed chemically resistant and inert
Suitable for dropping out aggressive chemicals.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 25 ml
    93 mm
    33 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 113,00 €
    113,00 €
    1 * 1 ST
       113,00 €
  • 50 ml
    100 mm
    43 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 125,00 €
    125,00 €
    1 * 1 ST
       125,00 €