


Iodine ≥99%, GPR RECTAPUR®, resublimed
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
:253.81 g/mol
Merck Index:13,05036
Boiling Pt:184.3 °C (1013 hPa)
Melting Pt:114 °C
Density:4.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-442-4
UN:3495
ADR:8 (6.1),III
REACH:01-2119485285-30
:GHS08
:GHS07
:GHS09
Specification Test Results
- Assay:Min. 99 %
- Non-volatile residue:Max. 0.05 %
- Cl + Br (as Cl):Max. 0.025 %
Specification Test Results
Related Information
Frequently Bought Together