


Iodine, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
:253.81 g/mol
Merck Index:13,05036
Boiling Pt:184.3 °C (1013 hPa)
Melting Pt:114 °C
Density:4.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-442-4
UN:3495
ADR:8,III
:GHS08
:GHS07
:GHS09
Specification Test Results
- Assay:min. 99.8 %
- Chloride and Bromine (as Cl):max. 0.003 %
- Non-volatile Matter:max. 0.01 %
Specification Test Results
Frequently Bought Together