Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Selenium
Selenium ≥99.5% (metals basis), powder -325 mesh
Selenium ≥99.5% (metals basis), powder -325 mesh
:
Thermo Fisher Scientific
Selenium ≥99.5% (metals basis), powder -325 mesh
:
Thermo Fisher Scientific
:
7782-49-2
:
:
Se
:
MFCD00134090
:
78.96 g/mol
:
12,08572
:
∼685 °C (1013 hPa)
:
217 °C
:
4.28 g/cm³ (20 °C)
:
Ambient
:
231-957-4
:
3288
:
6.1,III
:
GHS06
:
GHS08
msds_ALFA010601
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Ethanol, GPR RECTAPUR®
: VWR Chemicals
4-Methyl-2-pentanone ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Chloroform ≥98% stabilised, TECHNICAL
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Toluene, EMPLURA®, extra pure, Supelco®
: Merck
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
: Merck
Methanol, LiChrosolv®, hypergrade for LC-MS, for HPLC, Supelco®
: Merck
Acetone ≥99.0% Ph. Eur.
: VWR Chemicals
2-Propanol, SupraSolv® for gas chromatography, Supelco®
: Merck
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Formaldehyde ≥37% in aqueous solution stabilised, GR ACS, Reag. Ph. Eur. analytical reagent, Sigma-Aldrich®
: Merck
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Pyridine ≥99%, GPR RECTAPUR®
: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Ethanol absolute ≥99.9% (by GC), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Tetrahydrofuran stabilised, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck