ContactUsLinkComponent
96 square well, flat bottom plate, Falcon®
96 square well, flat bottom plate, Falcon®
Catalog # 734-0984
Supplier:  Corning
   Low Manufacturing Impact
undefined
96 square well, flat bottom plate, Falcon®
Catalog # 734-0984
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • No. of wells:
    96
  • Sterile:
    +
  • Description:
    96-square well plate, flat bottom
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford
  • Pk:
    5
  • Well volume:
    120 µl
Specifications
About this item
Sterile flat-bottom microplate with lid, made from crystal-grade virgin PS.
  • Sterilised by gamma irradiation
  • Non pyrogenic