Orders LinkContactUsLinkComponent
Funnels with short stem
Funnels with short stem
Supplier:   Brand
Funnels with short stem
Supplier:  Brand
Certificates
Funnels with short stem
Funnels with short stem
11 Options Available Below
Product Details & Documents
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With smooth internal walls. Flare angle 60°. Funnel lip is ground ore fire-polished.
Product Family Options
Product Information
  • Stem length
    Stem O-Ø
    Funnel O-Ø
    Availability
    Price
  • 35 mm
    6 mm
    35 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,20 €
    5,20 €
    1 * 1 ST
       5,20 €
  • 40 mm
    6 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,30 €
    5,30 €
    1 * 1 ST
       5,30 €
  • 50 mm
    7 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,00 €
    6,00 €
    1 * 1 ST
       6,00 €
  • 55 mm
    8 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,40 €
    6,40 €
    1 * 1 ST
       6,40 €
  • 60 mm
    8 mm
    60 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,90 €
    6,90 €
    1 * 1 ST
       6,90 €
  • 70 mm
    8 mm
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8,30 €
    8,30 €
    1 * 1 ST
       8,30 €
  • 80 mm
    9 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,10 €
    9,10 €
    1 * 1 ST
       9,10 €
  • 100 mm
    10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,60 €
    10,60 €
    1 * 1 ST
       10,60 €
  • 120 mm
    16 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,60 €
    38,60 €
    1 * 1 ST
       38,60 €
  • 150 mm
    16 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 55,30 €
    55,30 €
    1 * 1 ST
       55,30 €
  • 175 mm
    26 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 146,00 €
    146,00 €
    1 * 1 ST
       146,00 €