Orders LinkContactUsLinkComponent
Prefiltration capsules, Mini Profile®
Prefiltration capsules, Mini Profile®
Prefiltration capsules, Mini Profile®
Certificates
Prefiltration capsules, Mini Profile®
3 Options Available Below
Product Details & Documents
These are scaleable depth capsules for prefiltration and clarification. They are designed for small-volume production and scale-up evaluation.
  • High capacity depth medium is absolute particle rated for reliability
  • Broad chemical compatibility
  • Optimised for viscous fluids
  • Excellent for gel removal
Mini Profile II filters have a thick depth structure, providing high capacity for larger solids and gels as well as for fine particles.
Profile Star filters feature high area star-shaped pleated construction. The patented design combines the advantages of thick depth filters with those of traditional high area pleated filters.
PP filter media, core/end caps, and housing.
Specifications for Product Family
Specifications
  • Catalog No:
  • PALL12070
  • PALL12071
  • PALL12072
  • Type:
  • Profile Star filter, inlet/outlet hose barb Mini Profile®
  • Profile Star filter, inlet/outlet hose barb Mini Profile®
  • Profile Star filter, inlet/outlet hose barb Mini Profile®
  • Inlet/outlet:
  • 13 mm hose barb
  • 13 mm hose barb
  • 13 mm hose barb
  • Max. temperature:
  • 50 °C
  • 50 °C
  • 50 °C
  • Max. pressure:
  • 5,0 bar
  • 5,0 bar
  • 5,0 bar
Specifications
Product Family Options
Product Information
  • Type
    Pore size
    Filtration area
    Availability
    Price
  • Profile Star filter, inlet/outlet hose barb Mini Profile®
    1,5 µm
    90 cm²
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 354,00 €
    354,00 €
    1 * 3 ST
       354,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    PALL12070
    Type
    Profile Star filter, inlet/outlet hose barb Mini Profile®
    Inlet/outlet
    13 mm hose barb
    Max. temperature
    50 °C
    Max. pressure
    5,0 bar
  • Profile Star filter, inlet/outlet hose barb Mini Profile®
    3 µm
    90 cm²
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 354,00 €
    354,00 €
    1 * 3 ST
       354,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    PALL12071
    Type
    Profile Star filter, inlet/outlet hose barb Mini Profile®
    Inlet/outlet
    13 mm hose barb
    Max. temperature
    50 °C
    Max. pressure
    5,0 bar
  • Profile Star filter, inlet/outlet hose barb Mini Profile®
    5 µm
    90 cm²
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 354,00 €
    354,00 €
    1 * 3 ST
       354,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    PALL12072
    Type
    Profile Star filter, inlet/outlet hose barb Mini Profile®
    Inlet/outlet
    13 mm hose barb
    Max. temperature
    50 °C
    Max. pressure
    5,0 bar