ContactUsLinkComponent
Cell culture inserts, fibronectin, Corning® BioCoat™ FluoroBlok™
Cell culture inserts, fibronectin, Corning® BioCoat™ FluoroBlok™
  734-1053
 :  Corning
   Low Manufacturing Impact
undefined
Cell culture inserts, fibronectin, Corning® BioCoat™ FluoroBlok™
  734-1053
 :  Corning
   Low Manufacturing Impact
  • Membrane:
    PET
  • Pore size:
    3,0 µm
  • Description:
    Individual 3,0 µm inserts, ready to use in 24-well plate
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford
  • Pk:
    24
Corning® BioCoat™ FluoroBlok™ fibronectin-coated cell culture inserts packaged ready to use in 24-well Falcon® companion plates.
  • Convenient, individual inserts packaged in a one-piece, automation friendly 24-multiwell format
  • Combined benefits of Falcon® inserts coated with HFN for enhanced cell attachment and growth
  • Individual inserts contain a 3,0 µm pore size clear PET membrane