ContactUsLinkComponent
Racks for cryoboxes, Nunc™
Racks for cryoboxes, Nunc™
Catalog # 479-2483
undefined
Racks for cryoboxes, Nunc™
Catalog # 479-2483
Specifications
  • Colour code:
    Silver
  • Compartments:
    12
  • W×D×H ext.:
    143,2×140,4×670 mm
  • Pk:
    1
Specifications
About this item
Aluminium.
  • Fits most chest freezers
  • Adjustable shelf heights with a maximum of 12 boxes
  • Alignment rods secure boxes in position
  • Easy to clean, fits most chest freezers