Order Entry
Belgium
ContactUsLinkComponent
Orthophosphoric acid ≥85%
Corrosive
Orthophosphoric acid ≥85%
Catalog # VWRS6014.9190
Supplier:  VWR Collection
CAS Number:  
Corrosive
Orthophosphoric acid ≥85%
Catalog # VWRS6014.9190
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    176 L
  • Seq:
    5483

Specifications

About this item

 :
 :98 g/mol
Melting Pt:28 °C
Density:1.71…1.87 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
 :GHS05
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340