Disposable spill kit
Disposable spill kit
Catalog # 121-0136
Supplier: Brady
Jump to:
Product Details & Documents
Designed for small spills of chemicals and hazardous liquids up to 1,7 L. The highly visible dispenser box can easily be be wall or shelf mounted.
- Disposable brush to sweep up any broken glass or debris
- Protective gloves have roughened finger tips, increased tear resistance and good chemical resistance
- Autoclavable disposal bag
Delivery information: Kit contains two absorbent pads (410×510 mm), one pair of disposable chemical protective gloves (powder free), disposal bag, one disposable brush, and multilingual instruction guide.
Specifications
- Description:Disposable spill kit