Orders LinkContactUsLinkComponent
VWR® Basic Bouffant Caps, with Latex
VWR® Basic Bouffant Caps, with Latex
VWR® Basic Bouffant Caps, with Latex
Supplier:  VWR Collection
Certificates
VWR® Basic Bouffant Caps, with Latex
2 Options Available Below
Product Details & Documents
VWR® Basic bouffant caps are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Latex
  • Lightweight and breathable first line barrier protection
  • Tested and manufactured in an ISO certified facility that is lot-controlled and validated through independent laboratory testing
This garment is ideal for less critical areas or pre-gowning entry rooms.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-1461
  • 113-1462
  • Garment application:
  • Cleanroom
  • Cleanroom
Specifications
Product Family Options
Product Information
  • Colour
    Ø
    Availability
    Price
  • With latex
    White
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 74,40 €
    74,40 €
    1 * 1.000 ST
       74,40 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-1461
    Garment application
    Cleanroom
  • With latex
    White
    610 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 78,30 €
    78,30 €
    1 * 1.000 ST
       78,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-1462
    Garment application
    Cleanroom