Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, PBS tablets, pH 7,4, Millipore®

Buffer, PBS tablets, pH 7,4, Millipore®

Phosphate buffered saline (PBS) buffer Tablets, pH 7,4, 10 Tab.
Catalog # 524650-1
Buffer, PBS tablets, pH 7,4, Millipore®
Catalog # 524650-1
CAS Number:  
Buffer, PBS tablets, pH 7,4, Millipore®

Phosphate buffered saline (PBS) buffer Tablets, pH 7,4, 10 Tab.

CAS Number:  
Price
75,80 €
Log in for availability
1 * 1 EA
Product Details & Documents

Developed for use in immunoassay applications.

Dissolving one tablet of PBS buffer in 1 litre of deionised water yields 140 mM NaCl, 10 mM phosphate buffer, and 3 mM KCl, pH 7,4 at 25 °C.

Documents

Specifications

Specifications

  • Pk:
    10 Tab.
  • Description:
    Phosphate buffered saline (PBS) buffer Tablets, pH 7,4

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.