Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Pancreatin
Pancreatin
Pancreatin
Catalog # SERA31445.01
Supplier:
Serva
CAS Number:
Expand image
Pancreatin
Catalog # SERA31445.01
Supplier:
Serva
CAS Number:
Specifications
Pk:
1
CAS No.:
8049-47-6
Storage conditions:
Refrigerator
Enzyme name:
Pancreatin
Pk Info:
250 g
Specifications
About this item
Product Details
Product Details