Order Entry
Belgium
ContactUsLinkComponent
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form), Millipore®
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form), Millipore®
Catalog # 1.24541.0001
Supplier:  Merck
CAS Number:  
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form), Millipore®
Catalog # 1.24541.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :787.37 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Freezer
EINECS:246-129-8
CAS Number:24292-60-2
Formula:C₂₁H₂₆N₇Na₂O₁₇P₃
MDL Number:MFCD00065390