Order Entry
Belgium
ContactUsLinkComponent
 
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form) 97+%
 
undefined
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form) 97+%
 

 

  • Appearance:
    Yellow Powder
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥97.0 % (enzymatic, on dry subst.)
  • Water:
    ≤6.0 %
  • Solubility:
    (0.05 % in water) Clear colorless to light yellow

 

 

Frequently Bought Together