ContactUsLinkComponent
Methyl violet 2B, pure indicator grade
Irritant
Health
Environment
Methyl violet 2B, pure indicator grade
Certificates
undefined
Irritant
Health
Environment
Methyl violet 2B, pure indicator grade
Specification Test Results
  • Appearance:
    Green Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤10 % (105°C)
  • UV spectrophotometry:
    ≥75 % (on dry substance)
  • UV Lambda max:
    580 to 586 nm (in 50:50 IMS:water)
  • Absorption Ratio P-15/P+15:
    1 to 1.25