Order Entry
Belgium
ContactUsLinkComponent
Methyl violet 2B for microscopy, Sigma-Aldrich®
Corrosive
Health
Irritant
Environment
Methyl violet 2B for microscopy, Sigma-Aldrich®
Certificates
undefined
Corrosive
Health
Irritant
Environment
Methyl violet 2B for microscopy, Sigma-Aldrich®

Specification Test Results

  • Appearance (colour):
    Green to very dark green and brown to very dark brown and green-brown and brown-green
  • Appearance (form):
    Powder
  • Assay (gravimetr.):
    ≥80%
  • Thin layer chromatography:
    Corresponds to standard
  • Solubility (color):
    Red to very dark red
  • Solubility (method):
    0.01 g in 10ml water
  • Loss on drying:
    ≤7%
  • Infrared spectrum:
    Conforms to structure
  • Wavelength (1) (uv):
    509 - 524 nm
  • Extinction (1%/1cm) (1) (uv):
    ≥ 600
  • Solvent (uv):
    Water + 1 ml 1% sodium carbonate
  • Lead (Pb) by Element analysis:
    Not detected
  • Zinc (Zn) by Element analysis:
    Not detected
  • Colour:
    Violet
  • Extinction (1%/1 cm) (1) (uv):
    ≥ 1500

Specification Test Results

Frequently Bought Together