Lugol's iodine solution
Catalog # MSPP79012130.1000
Supplier: MARKET SOURCE PART PROCESS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient