Order Entry
Belgium
ContactUsLinkComponent
May Grunwald's eosin-methylene blue solution, Sigma-Aldrich®
Flammable
Toxic
Health
May Grunwald's eosin-methylene blue solution, Sigma-Aldrich®
Catalog # MG128-4L
CAS Number:  
Flammable
Toxic
Health
May Grunwald's eosin-methylene blue solution, Sigma-Aldrich®
Catalog # MG128-4L
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

About this item

 :
ADR:3,II
 :M-Clarity™ quality level = MQ500 (more info.)
UN:1230
 :GHS02
 :GHS06
 :GHS08
Storage Temperature:Ambient
 :1
MDL Number:MFCD00131580