Order Entry
Belgium
ContactUsLinkComponent
 
Irritant
May Grunwald's eosin-methylene blue for cell staining, Sigma-Aldrich®
Catalog # 205435-25G
CAS Number:  
Irritant
May Grunwald's eosin-methylene blue for cell staining, Sigma-Aldrich®
Catalog # 205435-25G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance (Colour):
    Green to Black
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • UV Absorbance Ratio
    A650/A523:
    ≥ 1.45

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD00131580