
Iron(II) sulphate hydrate, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:13463-43-9
:
Formula:FeSO₄·H₂O
MDL Number:MFCD00149722
:169.93 g/mol
Melting Pt:∼300 °C
Density:2.97 g/cm³ (20 °C)
EINECS:231-753-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together