Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cell Culture Supplements and Additives
Egg yolk emulsion
Egg yolk emulsion
Egg yolk emulsion
Catalog # OXOISR0047C
Supplier:
THERMO FISHER DIAGNOSTICS
CAS Number:
Expand image
Egg yolk emulsion
Catalog # OXOISR0047C
Supplier:
THERMO FISHER DIAGNOSTICS
CAS Number:
Specifications
Pk:
100 mL
Description:
Egg yolk emulsion
Specifications
About this item
Product Details
Product Details