Dextrin
Plastic bottle for solids, 250 g
Catalog # ROTH3488.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Dextrin
- CAS Number:9004-53-9
- Formula:(C₆H₁₂O₆)n
- MDL Number:MFCD00081554
- Molecular Weight:180.16 g/mol
- Merck Index:13,02971
- Density:1.45 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:232-675-4
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g



