Order Entry
Belgium
ContactUsLinkComponent
Folin-Ciocalteu's reagent for analysis of phenols
Irritant
Folin-Ciocalteu's reagent for analysis of phenols
Catalog # APPCA5084.0500
Supplier:  PanReac AppliChem
CAS Number:  
Irritant
Folin-Ciocalteu's reagent for analysis of phenols
Catalog # APPCA5084.0500
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
Density:1.22 g/cm³ (20 °C)
UN:3264
ADR:8,III
 :GHS07
MDL Number:MFCD00132625