0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Giemsa's stain
Giemsa's stain modified solution, Sigma-Aldrich®
Giemsa's stain modified solution, Sigma-Aldrich®
:
SIGMA ALDRICH MICROSCOPY
Giemsa's stain modified solution, Sigma-Aldrich®
:
SIGMA ALDRICH MICROSCOPY
:
51811-82-6
:
:
MFCD00081642
:
0.99 g/cm³ (20 °C)
:
1230
:
3,II
:
M-Clarity™ quality level = MQ200
:
11 °C
:
257-438-2
:
GHS02
:
GHS06
:
GHS08
:
Ambient
Frequently Bought Together
Test strips for peracetic acid, MQuant®, Supelco®
: Merck
Potassium hydroxide 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Test kits, phosphate, Spectroquant®, Supelco®
: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
: Merck
pH indicator strips, MQuant®, Supelco®
: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals