Order Entry
Belgium
Orders LinkContactUsLinkComponent
Docusate sodium 96%

Docusate sodium 96%

Docusate sodium 96%
Certificates
Docusate sodium 96%

3 Options Available Below

Product Details & Documents
Synonyms:  Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
  • CAS Number:577-11-7
  • Formula:C₂₀H₃₇NaO₇S
  • MDL Number:MFCD00012455
  • Molecular Weight:444.57 g/mol
  • Merck Index:12,03460
  • Boiling Pt:? 180 °C (1013 hPa)
  • Melting Pt:153…157 °C
  • Density:~ 1,1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:209-406-4

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Waxy solid
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥95 %
  • Water:
    ≤2 %
  • Color of alcohol solution:
    ≤100 APHA (50 % solids in 1:1 alcohol/water)
  • Insoluble matter:
    ≤300 ppm (50 % in toluene)
  • Iodine value:
    ≤0.25 (on solids basis)
  • Acid number:
    ≤2.5 (on solids basis)
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.