Docusate sodium ≥96%
About this item
Purity: 96% CAS No.: 577-11-7 Molecular Formula: C20H37NaO7S Formula Weight: 444.55 Physical Form: Solid Density: 1.1 Melting Point: 153-157° Solubility: Soluble in water, acetone, alcohol Merck Reference: 14,3401 MDL No.: MFCD00012455
Wetting and solubilizing agent
- CAS Number:577-11-7
- Formula:C₂₀H₃₇NaO₇S
- MDL Number:MFCD00012455
- Molecular Weight:444.57 g/mol
- Merck Index:14,3401
- Boiling Pt:∼180 °C (1013 hPa)
- Melting Pt:153…157 °C
- Density:1.1
- Storage Temperature:Ambient
- EINECS:209-406-4
2 Options Available Below
Product Details & Documents
Purity: 96% CAS No.: 577-11-7 Molecular Formula: C20H37NaO7S Formula Weight: 444.55 Physical Form: Solid Density: 1.1 Melting Point: 153-157° Solubility: Soluble in water, acetone, alcohol Merck Reference: 14,3401 MDL No.: MFCD00012455
Wetting and solubilizing agent
- CAS Number:577-11-7
- Formula:C₂₀H₃₇NaO₇S
- MDL Number:MFCD00012455
- Molecular Weight:444.57 g/mol
- Merck Index:14,3401
- Boiling Pt:∼180 °C (1013 hPa)
- Melting Pt:153…157 °C
- Density:1.1
- Storage Temperature:Ambient
- EINECS:209-406-4
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



