Quinine sulphate dihydrate
Catalog # MOLE29117911-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:782.96 g/mol
Merck Index:12,03822
Melting Pt:219 °C
Density:1.36 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-359-2
CAS Number:6119-70-6
:207671-44-1
Formula:C₄₀H₅₀N₄O₈S·2H₂O
MDL Number:MFCD00150790