ContactUsLinkComponent
Quinine sulphate dihydrate 99+%
Quinine sulphate dihydrate 99+%
Certificates
undefined
Quinine sulphate dihydrate 99+%
Certificates
Specification Test Results
  • Appearance:
    White to off-white Powder or crystals or needles
  • Infrared spectrum:
    Conforms
  • Water:
    4.0 to 5.5 % (K.F.)
  • Residue after ignition:
    ≤0.1 %
  • Specific optical rotation:
    -235° to -245° (20°C, 589 nm) (c=2, 0.1N HCl) (on anhydrous basis)
  • Titration with HClO4:
    99.0 to 101.0 % (On anhydrous basis)