Orders LinkContactUsLinkComponent
VWR®, Block Weights, Class M1
VWR®, Block Weights, Class M1
Supplier:   VWR Collection
VWR®, Block Weights, Class M1
Supplier:  VWR Collection
Certificates
VWR®, Block Weights, Class M1
VWR®, Block Weights, Class M1
8 Options Available Below
Product Details & Documents
Rectangular weights made from cast iron (density 7,2 g/cm³).
  • Indelible unique identification number guarantees traceability
Colour: Grey.
Conforms to OILM, COFRAC accredited
Certifications: Conforms to OIML, COFRAC accredited.
Packaging: Block weights are supplied with cardboard packaging to protect the weights during transportation.
Product Family Options
Product Information
  • Weight
    Tolerance
    Type
    Availability
    Price
  • 5 kg
    250 ± mg
    Without certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 104,00 €
    104,00 €
    1 * 1 ST
       104,00 €
  • 10 kg
    500 ± mg
    Without certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 140,00 €
    140,00 €
    1 * 1 ST
       140,00 €
  • 20 kg
    1000 ± mg
    Without certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 213,00 €
    213,00 €
    1 * 1 ST
       213,00 €
  • 50 kg
    2500 ± mg
    Without certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 533,00 €
    533,00 €
    1 * 1 ST
       533,00 €
  • 5 kg
    250 ± mg
    With certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 142,00 €
    142,00 €
    1 * 1 ST
       142,00 €
  • 10 kg
    500 ± mg
    With certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 177,00 €
    177,00 €
    1 * 1 ST
       177,00 €
  • 20 kg
    1000 ± mg
    With certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 231,00 €
    231,00 €
    1 * 1 ST
       231,00 €
  • 50 kg
    2500 ± mg
    With certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 575,00 €
    575,00 €
    1 * 1 ST
       575,00 €