ContactUsLinkComponent
Reverse osmosis systems, ReversinO
Reverse osmosis systems, ReversinO
Certificates
undefined
Reverse osmosis systems, ReversinO
Specifications
  • Catalog No:
  • 172-4216
  • 172-4215
  • Model:
  • ReversinO 6 Plus
  • ReversinO 3 Plus
  • Feed water:
  • Tap water
  • Tap water
  • Production flow rate:
  • 4 - 12 L/h
  • 2 - 6 L/h
  • W×D×H:
  • 370×250×700 mm
  • 370×250×700 mm
  • Weight:
  • 12 kg
  • 11,5 kg