Orders LinkContactUsLinkComponent
Boiling baths, SBB Aqua Plus series
Boiling baths, SBB Aqua Plus series
Supplier:   GRANT INSTRUMENTS
Boiling baths, SBB Aqua Plus series
Supplier:  GRANT INSTRUMENTS
Certificates
Boiling baths, SBB Aqua Plus series
Boiling baths, SBB Aqua Plus series
3 Options Available Below
Product Details & Documents
Boiling baths are designed for continuous operation with water above 90 °C. They have a stainless steel tank in an outer case and analogue control by means of a simple dial for straightforward temperature settings. Baths have two fixed over-temperature safety cut-outs which can be reset by the user.
  • Energy regulator can be adjusted to ensure a steady boiling action, avoiding excessive steam or spitting
  • Constant level device maintains the required liquid level, avoiding the need to monitor and top up the bath
  • Heater and temperature sensors are mounted at the base of the tank, protected by a perforated tray, giving more usable space
  • Robust construction, durable and resistant to corrosion
  • New modern design
Required accessories must be ordered separately, see SUB Aqua Plus baths entry.
Ordering information: Required accessories must be ordered separately, see SUB Aqua Pro baths entry.
Delivery information: Supplied with polycarbonate lid but without rings.
Caution: ** Required if flat-bottomed flasks are to be placed directly onto the base of the bath and to allow better convection in the bath.
Product Family Options
Product Information
  • Type
    Temp.
    Heating capacity
    Capacity
    Bath W×D×H
    W×D×H
    Availability
    Price
  • SBB Aqua 5 Plus
    100 °C
    1,3/1,5 kW
    5 L
    290×145×125 mm
    390×250×375 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 370,00 €
    1 370,00 €
    1 * 1 ST
       1 370,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    462-0171
  • SBB Aqua 12 Plus
    100 °C
    1,35/1,5 kW
    12 L
    290×315×125 mm
    390×385×440 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 490,00 €
    1 490,00 €
    1 * 1 ST
       1 490,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    462-0166
  • SBB Aqua 26 Plus
    100 °C
    1,35/2,0 kW
    26 L
    290×145×175 mm
    390×600×445 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 850,00 €
    1 850,00 €
    1 * 1 ST
       1 850,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    462-0170
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel rack for 1.5 ml microtubes
    Stainless steel rack for 1.5 ml microtubes
    462-8458
    65 tubes
    Log in for availability
    1 * 1 ST - 150,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel rack for 0.5 ml microtubes
    Stainless steel rack for 0.5 ml microtubes
    462-8457
    105 tubes
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel base tray**
    Stainless steel base tray**
    462-8350
    SAP12, JBN12, JBA12, GLS Aqua 12 Plus , SBB Aqua 12 Plus
    Log in for availability
    1 * 1 ST - 116,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel base tray**
    Stainless steel base tray**
    462-8351
    SAP18/26. JBN18/26, JBA18, GLS Aqua 18 Plus, OLS200 , SBB Aqua 18/26 Plus
    Log in for availability
    1 * 1 ST - 123,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel test tube rack for 30 mm Ø tubes
    Stainless steel test tube rack for 30 mm Ø tubes
    462-8456
    12 tubes
    Log in for availability
    1 * 1 ST - 150,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel test tube rack for 19 mm Ø tubes
    Stainless steel test tube rack for 19 mm Ø tubes
    462-8454
    32 tubes
    Log in for availability
    1 * 1 ST - 150,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel test tube rack for 25 mm Ø tubes
    Stainless steel test tube rack for 25 mm Ø tubes
    462-8455
    18 tubes
    Log in for availability
    1 * 1 ST - 150,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel test tube rack for 16 mm Ø tubes
    Stainless steel test tube rack for 16 mm Ø tubes
    462-8453
    36 tubes
    Log in for availability
    1 * 1 ST - 150,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel test tube rack for 13 mm Ø tubes
    Stainless steel test tube rack for 13 mm Ø tubes
    462-8452
    55 tubes
    Log in for availability
    1 * 1 ST - 150,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel test tube rack for 10 mm Ø tubes
    Stainless steel test tube rack for 10 mm Ø tubes
    462-8451
    84 tubes
    Log in for availability
    1 * 1 ST - 150,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Gabled stainless steel lid
    Gabled stainless steel lid
    462-8212
    JBA18, JBN18/26, SAP18/26, (SUB Aqua 18/26, JB Aqua 18/26, SBB Aqua 18/26, GLS Aqua 18 Plus), LSB18
    Log in for availability
    1 * 1 ST - 296,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Gabled stainless steel lid
    Gabled stainless steel lid
    462-8211
    JBA12, JBN12, SAP12, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus, GLS Aqua 12 Plus), LSB12
    Log in for availability
    1 * 1 ST - 264,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Gabled stainless steel lid
    Gabled stainless steel lid
    462-8210
    JBA5, JBN5, SAP2S, SAP5, (SUB Aqua 5, SUB Aqua Dual, JB Aqua 5, JB Aqua Dual, SBB Aqua 5 Plus)
    Log in for availability
    1 * 1 ST - 240,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Flat lid with 6 ring sets
    Flat lid with 6 ring sets
    462-8222
    JBA18, JBN18/26, SAP18/26, (SUB Aqua 18/26, JB Aqua 18/26, SBB Aqua 18/26 Plus)
    Log in for availability
    1 * 1 ST - 611,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Flat lid with 4 ring sets
    Flat lid with 4 ring sets
    462-8221
    JBA12, JBN12, SAP12, SAPD, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus)
    Log in for availability
    1 * 1 ST - 440,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Raised half shelf with perforated mesh
    Raised half shelf with perforated mesh
    462-8232
    JBN26, SAP26, (SUB Aqua 26, JB Aqua 26, SBB Aqua 26 Plus)
    Log in for availability
    1 * 1 ST - 255,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Flat lid with 2 ring sets
    Flat lid with 2 ring sets
    462-8220
    JBA5, JBN5, SAP5, SAPD, (SUB Aqua 5, SUB Aqua Dual, JB Aqua 5, JB Aqua Dual, SBB Aqua 5 Plus)
    Log in for availability
    1 * 1 ST - 286,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Raised half shelf with perforated mesh
    Raised half shelf with perforated mesh
    462-0187
    JBA18, JBN18, SAP18, (SUB Aqua 18, JB Aqua 18, SBB Aqua 18 Plus)
    Log in for availability
    1 * 1 ST - 215,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Raised half shelf with perforated mesh
    Raised half shelf with perforated mesh
    462-8231
    JBA12, JBN12, SAP12, SAPD, (SUB Aqua 12, SUB Aqua Dual, JB Aqua 12, JB Aqua Dual, SBB Aqua 12 Plus)
    Log in for availability
    1 * 1 ST - 182,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement lids, polycarbonate
    Replacement lids, polycarbonate
    GRANAQL5
    SAP2S, SAP5 , JBN5, JBA5, SBB Aqua 5 Plus
    Log in for availability
    1 * 1 ST - 122,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Polycarbonate lid
    Polycarbonate lid
    462-0333
    Log in for availability
    1 * 1 ST - 150,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Polycarbonate lid
    Polycarbonate lid
    462-0334
    Log in for availability
    1 * 1 ST - 171,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Stainless steel base tray**, for SAP2S, SAP5, JBN5, JBA5 and SBB Aqua 5 Plus
    Stainless steel base tray**, for SAP2S, SAP5, JBN5, JBA5 and SBB Aqua 5 Plus
    462-8131
    Log in for availability
    1 * 1 ST - 123,00 €
Product Information
  • Catalog #
    Material
    Ø
    Availability
    Price
  • Anti-evaporation spheres
    Anti-evaporation spheres
    462-8239
    PP
    20 mm
    Log in for availability
    1 * 300 ST - 123,00 €