Water bath protective media, Aqua-Stabil
Leverancier: JULABO GmbH
Certificaten
Over dit artikel
Ideal for use in heating baths, circulators, water baths and shaking water baths. Use 2 ml solution per 1 litre water.
- Prevents contamination and the formation of algae, bacteria and microorganisms
Let op: Use biocides safely. Always read the label and product information before use.
Frequently Bought Together