Orders LinkContactUsLinkComponent
Water baths, aqualine series
Water baths, aqualine series
Supplier:  
Certificates
Water baths, aqualine series
Water baths, aqualine series
0 Options Available Below
Product Details & Documents
Unstirred, corrosion resistant stainless steel baths, with heater and temperature sensors mounted underneath the tank ensuring maximum usable space. Robust and reliable, controls are recessed for protection, baths are easy to clean and the minimum filling Level is just 20 mm.
  • Easy operation via digital LED display
  • Integrated over-temperature cut-out
  • Angled lid prevents condensation dripping into samples
  • Homogenous temperature distribution thanks to heating elements positioned underneath the tank
Temperature stability at 37 °C: ±0,2 K.
Temperature stability at 37 °C (K): ±0,2
 : IP protection class according to DIN EN 60529: IP 22.
 : Supplied with a transparent polycarbonate gabled lid, but without racks or platform, required accessories must be ordered separately.
 : D = overall depth (open lid + controller), H = with lid closed.
Product Family Options
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • Stainless steel lifting platform
    Stainless steel lifting platform
    462-1018
    AL 12 (max. 1 platform), AL 18 and AL 25 (max. 2 platforms)
    Log in for availability
    1 * 1 ST - 488,00 €
Product Information
  • Catalog #
    No. of Holes
    For tube Ø
    Availability
    Price
  • Stainless steel rack for 1×in AL 5, 3×in AL 12, 5×in AL 18, AL 25
    Stainless steel rack for 1×in AL 5, 3×in AL 12, 5×in AL 18, AL 25
    462-1009
    21
    14/18 mm
    Log in for availability
    1 * 1 ST - 308,00 €
Product Information
  • Catalog #
    No. of Holes
    For tube Ø
    Availability
    Price
  • Stainless steel rack for 1×in AL 5, 3×in AL 12, 5×in AL 18, AL 25
    Stainless steel rack for 1×in AL 5, 3×in AL 12, 5×in AL 18, AL 25
    462-1010
    21
    14/18 mm
    Log in for availability
    1 * 1 ST - 308,00 €
Product Information
  • Catalog #
    No. of Holes
    For tube Ø
    Availability
    Price
  • Stainless steel rack for 1×in AL 5, 3×in AL 12, 5×in AL 18, AL 25
    Stainless steel rack for 1×in AL 5, 3×in AL 12, 5×in AL 18, AL 25
    462-1011
    10
    24/30 mm
    Log in for availability
    1 * 1 ST - 308,00 €
Product Information
  • Catalog #
    No. of Holes
    For tube Ø
    Availability
    Price
  • Stainless steel rack for 2×in AL 5
    Stainless steel rack for 2×in AL 5
    462-1002
    11
    14/18 mm
    Log in for availability
    1 * 1 ST - 316,00 €
Product Information
  • Catalog #
    No. of Holes
    For tube Ø
    Availability
    Price
  • Stainless steel rack for 1×in AL 12, 2×in AL 18, AL 25
    Stainless steel rack for 1×in AL 12, 2×in AL 18, AL 25
    462-1007
    15
    24/30 mm
    Log in for availability
    1 * 1 ST - 516,00 €