Orders LinkContactUsLinkComponent
Vacuum Controller, V-850/V-855
Vacuum Controller, V-850/V-855
Fournisseur:   BUCHI
Vacuum Controller, V-850/V-855
Fournisseur:  BUCHI
Certificates
Vacuum Controller, V-850/V-855
Vacuum Controller, V-850/V-855
2 Options Available Below
Product Details & Documents
V-850 vacuum controller:
The V-850 vacuum controller with ¼" LCD VGA screen can be used universally for the display, control and adjustment of vacuum. The vacuum controller is a compact unit suitable for many tasks in the laboratory thanks to its integrated vent valve and chemical-resistant pressure sensor. It includes a timer, laboratory vacuum controller (Labvac) and solvent database featuring the 43 most commonly used solvents. The rotation speed control with the V-700 or V-710 vacuum pump enables accurate, hysteresis-free vacuum control with quiet operation.
Users also have the option of reducing their water consumption by adding a cooling water valve.
V-855 vacuum controller:
The V-855 vacuum controller comes with the same features as the V-850, and in addition: gradient and repetition functions for distillation in difficult conditions and the EasyVac function — automatic distillation based on vapour pressure detection.
The V-855 vacuum controller comes with the same features as the V-850, and in addition: gradient and repetition functions for distillation in difficult conditions and the EasyVac function — automatic distillation based on vapour pressure detection.
Delivery information: Delivered with an air vent valve and pressure sensors.
Product Family Options
Product Information
  • Disponibilité
    Prix
  • 531-0234
    Vacuum Controller V-855 for R-210/R-215
    Vacuum Controller V-855 for R-210/R-215
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 531-0232
    Vacuum Controller V-850 for R-210/R-215
    Vacuum Controller V-850 for R-210/R-215
    Arrêté
  • 181-0140
    Valve unit for V-850/V-855
    Valve unit for V-850/V-855
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 742,00 €
    742,00 €
    1 * 1 ST
       742,00 €
Accessories
Product Information
  • N° de catalogue
    Disponibilité
    Prix
  • Communication cable for R-210/R-215
    Communication cable for R-210/R-215
    181-0138
    Log in for availability
    1 * 1 ST - 34,90 €
Product Information
  • N° de catalogue
    Disponibilité
    Prix
  • Woulff bottle for V-850/V-855
    Woulff bottle for V-850/V-855
    181-0141
    Log in for availability
    1 * 1 ST - 237,00 €
Product Information
  • N° de catalogue
    Disponibilité
    Prix
  • Automatic distillation probe for V-855
    Automatic distillation probe for V-855
    531-0181
    Log in for availability
    1 * 1 ST - 250,00 €
Product Information
  • N° de catalogue
    Disponibilité
    Prix
  • Assembly set for R-210/R-215
    Assembly set for R-210/R-215
    531-0236
    Log in for availability
    1 * 1 ST - 76,80 €
Product Information
  • N° de catalogue
    Disponibilité
    Prix
  • Remote control for V-850/V-855
    Remote control for V-850/V-855
    531-0237
    Log in for availability
    1 * 1 ST - 669,00 €