Orders LinkContactUsLinkComponent
Thermometer test caps
Thermometer test caps
Supplier:   COMARK
Thermometer test caps
Supplier:  COMARK
Certificates
Thermometer test caps
Thermometer test caps
4 Options Available Below
Product Details & Documents
Certified test capsules can be used for quick checking of the thermometer's function against specifications. They are connected to the thermometer instead of a sensor and consist of a resistor that corresponds exactly to a given temperature.
Product Family Options
Product Information
  • Range
    Availability
    Price
  • 620-0377
    TX23L, test cap, Lumberg type
    TX23L, test cap, Lumberg type
    0 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
  • 620-0378
    TX24L, test cap, Lumberg type
    TX24L, test cap, Lumberg type
    +3 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
  • 620-0379
    TX25L, test cap, Lumberg type
    TX25L, test cap, Lumberg type
    +63 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
  • 620-0380
    TX26L, test cap, Lumberg type
    TX26L, test cap, Lumberg type
    +70 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €