Stirrer Guides with Anti-clockwise Thread
Supplier: Lenz Laborglas
Jump to:
About this item
DURAN®, borosilicate glass, PFTE/fibre glass pressure screw. The fluoroplastic bearing is in a borosilicate glass housing, and has adjustable PTFE/FPM seal. All parts are interchangeable.
2 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, PFTE/fibre glass pressure screw. The fluoroplastic bearing is in a borosilicate glass housing, and has adjustable PTFE/FPM seal. All parts are interchangeable.
Product Family Options
Product Information
- NSShaft ØAvailabilityPrice
- LENZ06306004Stirrer guide, dynamic double seal
- LENZ06306204Stirrer guide, dynamic double seal