Accessories for Spectroquant® NOVA
: Merck
To document the analysis values, either a standard computer or a printer can be connected.
- The Multi-Agate II software can control data acquisition or simply record recorded analysis data
- The data records are archived here as raw data or imported into further programs such as Excel
Connectable photometers: SQ 118, SQ 200, SQ 300, Spectroquant® NOVA 30, Spectroquant® NOVA 60, Spectroquant® NOVA 400.
To document the analysis values, either a standard computer or a printer can be connected.
- The Multi-Agate II software can control data acquisition or simply record recorded analysis data
- The data records are archived here as raw data or imported into further programs such as Excel
Connectable photometers: SQ 118, SQ 200, SQ 300, Spectroquant® NOVA 30, Spectroquant® NOVA 60, Spectroquant® NOVA 400.
Product Family Options
- DescriptionFor
- Software for Windows (in English and German) for the data transferNOVA photometers Spectroquant®1 * 1 ST
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.