Accessories for pHotoFlex®
: WTW
Frequently Bought Together
Accessories for VWR® V-1200, UV-1600PC, UV-3100PC
: VWR Collection
Objectives EC Epiplan
: ZEISS
UV/Vis Calibration Standards
: HELLMA
VWR®, Accessories for PerfectBlue Sedec Semi-Dry Blotters
: VWR Collection
PEEKLOK 0.36MM -1/32 FERRULE PK10 1 * 10 items
: Trajan Scientific and Medical
with Multi-Axis-stand (min. additional need : Lens and Monitor) 1 * 1 items
: VISION ENGINEERING
Gas detection system, X-am® 5000
: Dräger
Multi-parameter meter, handheld, SevenGo Duo pro™ SG98
: Mettler - Toledo
Evolution™ UV/VIS Spectrophotometers
: Thermo Fisher Scientific
Cuvettes for PFX190 series
: Lovibond Tintometer
INSTRUMENT CAS
: FINE SCIENCE TOOLS
Accessories for fibre determination systems
: GERHARDT
WATERBATH WTB6, 7.5L, 10-100C 1 * 1 Pce
: MARKET SOURCE PART PROCESS