3 Dimensional platform rotator, PS-3D
PS-3D platform rotator
Catalog # 444-0350
Supplier: GRANT INSTRUMENTS
Jump to:
Product Details & Documents
The compact PS-3D variable speed, fixed angle 3D rotator is ideally suited for personal use. Applications include mixing in tubes, culture flasks, gel staining, destaining, antibody staining, washes, hybridisations, and blots. The reliable motor produces regulated and reproducible rotating throughout the speed range.
- Gentle movement keeps fragile tissue intact during incubations
- Can be used at 4 to 40 °C (80% relative humidity, non-condensing), in an incubator or cold room
Delivery information: Supplied with non-slip mat, optional dimpled mat is available separately.
Documents
Specifications
- Description:PS-3D platform rotator
- Shaking motion:3D Orbital
- Cat. No.:444-0350
- Speed:5 - 60 min⁻¹
- Input power:External power supply: 12 V, 500 mA
- Angle:7°
- Model:PS-3D
- W×D×H:85×150×80 mm
- Max. load:0,5 kg
- Platform W×D:200×200 mm
- Weight:1,3 kg
Accessories
Product Information
- Catalog #AvailabilityPrice
- Dimpled mat
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



