ContactUsLinkComponent
Microplate shakers, MicroPlate Genie® and Multi-MicroPlate Genie®
undefined
Microplate shakers, MicroPlate Genie® and Multi-MicroPlate Genie®
  • Catalog No:
  • 444-0875
  • Model:
  • Digital MicroPlate Genie®
  • Speed range:
  • 500 - 2850 min⁻¹
  • Orbit:
  • 1,0 mm
  • No. of microplates held:
  • 1
  • Max. load:
  • 0,5 kg
  • Weight:
  • 2,75 kg
  • W×D×H:
  • 180×150×165 mm
  • Timer:
  • 1 - 99 min/continuous
Frequently Bought Together