Orders LinkContactUsLinkComponent
Ice sampler
Ice sampler
Supplier:   BURKLE
Ice sampler
Supplier:  BURKLE
Certificates
Ice sampler
3 Options Available Below
Product Details & Documents
Sampler developed for sampling from frozen, highly viscous, paste-like materials such as ice-cream and similar substances. The drill-shaped cutter screws itself into the frozen goods independently when being rotated. The collection cylinder can easily be detached via a bayonet connection. The sample is then available and can be removed.
  • Stainless steel AISI 316L (1.4404), electro-polished
  • Sterilisable
  • Easy cleaning
  • Collection cylinder 38 mm Ø
Product Family Options
Product Information
  • Length
    Ø
    Volume
    Availability
    Price
  • 300-1049
    Ice sampler complete
    Ice sampler complete
    1050 mm
    38 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 856,00 €
    856,00 €
    1 * 1 ST
       856,00 €
  • 300-1047
    Ice sampler complete
    Ice sampler complete
    300 mm
    38 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 757,00 €
    757,00 €
    1 * 1 ST
       757,00 €
  • 300-1048
    Ice sampler complete
    Ice sampler complete
    550 mm
    38 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 807,00 €
    807,00 €
    1 * 1 ST
       807,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Transport case
    Transport case
    300-1050
    Log in for availability
    1 * 1 ST - 24,70 €